2,2,2-Trifluoro-1-(2-fluoro-3-(methylthio)phenyl)ethanone ≥97%
Supplier: AOBChem USA
|
Warning
|
64460-500MG
64460-1G
64460-5G
64460-25G
77332-210EA
251.32
USD
77332-210
77356-568
77368-280
77389-588
2,2,2-Trifluoro-1-(2-fluoro-3-(methylthio)phenyl)ethanone ≥97%
2,2,2-Trifluoro-1-(2-fluoro-3-(methylthio)phenyl)ethanone
Precaución: for Research Use Only
Formula:
C₉H₆F₄OS MW: 238.2019 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD26687738 CAS Number: 1515166-46-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...