(5-Bromo-2-fluoro-3,4-dimethylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

64440-500MG 64440-1G 64440-5G 64440-25G
77332-204EA 98.88 USD
77332-204 77342-726 77368-276 77380-500
(5-Bromo-2-fluoro-3,4-dimethylphenyl)(methyl)sulfane ≥97%
(5-Bromo-2-fluoro-3,4-dimethylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₉H₁₀BrFS
Storage Temperature: Ambient
MDL Number: MFCD34761771
CAS Number: 2807454-24-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR