5-Bromo-2-fluoro-3,4-dimethylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

64435-500MG 64435-1G 64435-5G 64435-25G
77332-202EA 116.39 USD
77332-202 77343-438 77368-274 77380-498
5-Bromo-2-fluoro-3,4-dimethylbenzoic acid ≥97%
5-Bromo-2-fluoro-3,4-dimethylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₈BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD34761767
CAS Number: 2133041-58-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR