4-Fluor-2,3-dimethylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

64427-500MG 64427-1G 64427-5G 64427-25G
77332-198EA 90.64 USD
77332-198 77342-400 77368-270 77380-496
4-Fluor-2,3-dimethylbenzaldehyde ≥97%
4-Fluor-2,3-dimethylbenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉FO
Storage Temperature: Ambient
MDL Number: MFCD16295095
CAS Number: 363134-37-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR