1,5-Dichloro-3-fluoro-2-isopropoxybenzene ≥95%

Supplier: AOBChem USA

Warning

64422-500MG 64422-1G 64422-5G 64422-25G
77332-196EA 192.61 USD
77332-196 77356-562 77368-268 77389-582
1,5-Dichloro-3-fluoro-2-isopropoxybenzene ≥95%
1,5-Dichloro-3-fluoro-2-isopropoxybenzene

Precaución: for Research Use Only

Formula: C₉H₉Cl₂FO
MW: 223.0715 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29044158
CAS Number: 1881293-55-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR