2-(Benzyloxy)-1,5-dichloro-3-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

64421-500MG 64421-1G 64421-5G 64421-25G
77332-194EA 187.46 USD
77332-194 77356-560 77368-266 77389-580
2-(Benzyloxy)-1,5-dichloro-3-fluorobenzene ≥97%
2-(Benzyloxy)-1,5-dichloro-3-fluorobenzene

Precaución: for Research Use Only

Formula: C₁₃H₉Cl₂FO
MW: 271.1143 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29044321
CAS Number: 1881327-93-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR