1,5-Dichloro-2-ethoxy-3-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

64420-500MG 64420-1G 64420-5G 64420-25G
77332-192EA 234.84 USD
77332-192 77356-558 77368-264 77389-578
1,5-Dichloro-2-ethoxy-3-fluorobenzene ≥97%
1,5-Dichloro-2-ethoxy-3-fluorobenzene

Precaución: for Research Use Only

Formula: C₈H₇Cl₂FO
MW: 209.0449 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28785826
CAS Number: 1805126-73-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR