4-Isobutoxy-1-naphthaldehyde ≥97%

Supplier: AOBChem USA

Warning

64387-500MG 64387-1G 64387-5G
77338-750EA 513.97 USD
77338-750 77356-556 77380-486
4-Isobutoxy-1-naphthaldehyde ≥97%
4-Isobutoxy-1-naphthaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₆O₂
MW: 228.2863 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03928893
CAS Number: 700858-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR