4-Isobutoxy-1-naphthoic acid ≥97%

Supplier: AOBChem USA

Warning

64385-500MG 64385-1G 64385-5G
77338-430EA 487.19 USD
77338-430 77356-554 77380-484
4-Isobutoxy-1-naphthoic acid ≥97%
4-Isobutoxy-1-naphthoic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₆O₃
MW: 244.2857 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27966496
CAS Number: 1242811-15-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR