1-Ethoxy-4-iodonaphthalene ≥95%

Supplier: AOBChem USA

Warning

64380-500MG 64380-1G 64380-5G 64380-25G
77332-190EA 207.03 USD
77332-190 77356-552 77368-252 77389-576
1-Ethoxy-4-iodonaphthalene ≥95%
1-Ethoxy-4-iodonaphthalene

Precaución: for Research Use Only

Formula: C₁₂H₁₁IO
Storage Temperature: Ambient
MDL Number: MFCD17012468
CAS Number: 104296-74-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR