(4-Ethoxynaphthalen-1-yl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

64379-500MG 64379-1G 64379-5G 64379-25G
77332-188EA 152.44 USD
77332-188 77345-498 77368-250 77389-574
(4-Ethoxynaphthalen-1-yl)(methyl)sulfane ≥97%
(4-Ethoxynaphthalen-1-yl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₃H₁₄OS
MW: 218.3146 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761745
CAS Number: 2901098-56-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR