4-Ethoxy-1-naphthaldehyde ≥97%

Supplier: AOBChem USA

Warning

64377-500MG 64377-1G 64377-5G 64377-25G
77332-186EA 151.41 USD
77332-186 77345-402 77368-248 77389-572
4-Ethoxy-1-naphthaldehyde ≥97%
4-Ethoxy-1-naphthaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₁₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00859279
CAS Number: 54784-08-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR