4-Bromo-4'-methoxy-3-(trifluoromethyl)-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

64368-500MG 64368-1G 64368-5G 64368-25G
77332-180EA 204.97 USD
77332-180 77356-548 77368-242 77389-566
4-Bromo-4'-methoxy-3-(trifluoromethyl)-1,1'-biphenyl ≥95%
4-Bromo-4'-methoxy-3-(trifluoromethyl)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₀BrF₃O
MW: 331.1278 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34642048
CAS Number: 2807441-84-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR