4-Bromo-4'-fluoro-3-(trifluoromethyl)-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

64367-500MG 64367-1G 64367-5G 64367-25G
77332-178EA 233.81 USD
77332-178 77356-546 77368-240 77389-564
4-Bromo-4'-fluoro-3-(trifluoromethyl)-1,1'-biphenyl ≥97%
4-Bromo-4'-fluoro-3-(trifluoromethyl)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₇BrF₄
MW: 319.0922 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761742
CAS Number: 1343404-90-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR