4'-Bromo-2-methyl-3'-(trifluoromethyl)-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

64366-500MG 64366-1G 64366-5G 64366-25G
77332-176EA 204.97 USD
77332-176 77356-544 77368-238 77389-562
4'-Bromo-2-methyl-3'-(trifluoromethyl)-1,1'-biphenyl ≥97%
4'-Bromo-2-methyl-3'-(trifluoromethyl)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₀BrF₃
MW: 315.1284 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761741
CAS Number: 1343404-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR