4-Bromo-4'-methyl-3-(trifluoromethyl)-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

64365-500MG 64365-1G 64365-5G 64365-25G
77332-174EA 169.95 USD
77332-174 77346-362 77368-236 77389-560
4-Bromo-4'-methyl-3-(trifluoromethyl)-1,1'-biphenyl ≥95%
4-Bromo-4'-methyl-3-(trifluoromethyl)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₀BrF₃
MW: 315.1284 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761740
CAS Number: 1186482-67-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR