4-Bromo-5-fluoro-2,2'-dimethyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

64359-500MG 64359-1G 64359-5G 64359-25G
77332-164EA 139.05 USD
77332-164 77344-676 77368-230 77389-552
4-Bromo-5-fluoro-2,2'-dimethyl-1,1'-biphenyl ≥97%
4-Bromo-5-fluoro-2,2'-dimethyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₂BrF
MW: 279.1474 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761734
CAS Number: 2807441-70-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR