4-Bromo-5-fluoro-2,4'-dimethyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

64358-500MG 64358-1G 64358-5G 64358-25G
77332-162EA 169.95 USD
77332-162 77346-360 77368-228 77389-550
4-Bromo-5-fluoro-2,4'-dimethyl-1,1'-biphenyl ≥97%
4-Bromo-5-fluoro-2,4'-dimethyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₂BrF
MW: 279.1474 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761733
CAS Number: 2807454-40-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR