3-(4-Bromo-3-methoxyphenyl)pyridine ≥97%
Supplier: AOBChem USA
|
Warning
|
64354-500MG
64354-1G
64354-5G
77339-022EA
544.87
USD
77339-022
77356-536
77380-476
3-(4-Bromo-3-methoxyphenyl)pyridine ≥97%
3-(4-Bromo-3-methoxyphenyl)pyridine
Precaución: for Research Use Only
Formula:
C₁₂H₁₀BrNO MW: 264.1179 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34642120 CAS Number: 2807449-98-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...