3-(Benzyloxy)-2,4-difluoro-5-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

64338-500MG 64338-1G 64338-25G 64338-5G
77332-156EA 297.67 USD
77332-156 77356-534 77389-544 77380-468
3-(Benzyloxy)-2,4-difluoro-5-methylbenzaldehyde ≥97%
3-(Benzyloxy)-2,4-difluoro-5-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₂F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD34761725
CAS Number: 947279-26-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR