3-(Benzyloxy)-2,4-difluoro-5-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

64336-500MG 64336-1G 64336-5G 64336-25G
77332-154EA 264.71 USD
77332-154 77356-532 77368-220 77389-542
3-(Benzyloxy)-2,4-difluoro-5-methylbenzoic acid ≥97%
3-(Benzyloxy)-2,4-difluoro-5-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₂F₂O₃
MW: 278.2507 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761723
CAS Number: 2807449-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR