1-(6-Fluoro-2-methoxy-3-methylphenyl)ethanol ≥97%

Supplier: AOBChem USA

Warning

64327-500MG 64327-1G 64327-5G 64327-25G
77332-152EA 280.16 USD
77332-152 77356-528 77368-218 77389-540
1-(6-Fluoro-2-methoxy-3-methylphenyl)ethanol ≥97%
1-(6-Fluoro-2-methoxy-3-methylphenyl)ethanol

Precaución: for Research Use Only

Formula: C₁₀H₁₃FO₂
Storage Temperature: Ambient
CAS Number: 1785558-36-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR