3-Acetyl-2-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

64311-500MG 64311-1G 64311-5G 64311-25G
77332-150EA 219.39 USD
77332-150 77356-526 77368-216 77389-538
3-Acetyl-2-fluorobenzaldehyde ≥97%
3-Acetyl-2-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₇FO₂
Storage Temperature: Ambient
MDL Number: MFCD34761715
CAS Number: 2385435-38-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR