1-(2-Fluoro-3-(methylthio)phenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

64305-500MG 64305-1G 64305-5G 64305-25G
77332-148EA 204.97 USD
77332-148 77356-524 77368-214 77389-536
1-(2-Fluoro-3-(methylthio)phenyl)ethanone ≥97%
1-(2-Fluoro-3-(methylthio)phenyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₉FOS
MW: 184.2305 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26687728
CAS Number: 1504447-19-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR