3-Acetyl-2-fluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

64303-500MG 64303-1G 64303-5G 64303-25G
77332-146EA 188.49 USD
77332-146 77356-522 77368-212 77389-534
3-Acetyl-2-fluorobenzoic acid ≥97%
3-Acetyl-2-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₉H₇FO₃
MW: 182.1484 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26687988
CAS Number: 1508190-04-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR