1-(Benzyloxy)-2-bromo-4,5-dichlorobenzene ≥97%

Supplier: AOBChem USA

Warning

64301-500MG 64301-1G 64301-5G 64301-25G
77332-144EA 120.51 USD
77332-144 77343-780 77368-210 77380-464
1-(Benzyloxy)-2-bromo-4,5-dichlorobenzene ≥97%
1-(Benzyloxy)-2-bromo-4,5-dichlorobenzene

Precaución: for Research Use Only

Formula: C₁₃H₉BrCl₂O
MW: 332.0199 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761711
CAS Number: 2807455-72-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR