1-Bromo-4,5-dichloro-2-isopropoxybenzene ≥97%

Supplier: AOBChem USA

Warning

64294-500MG 64294-1G 64294-5G 64294-25G
77332-140EA 138.02 USD
77332-140 77344-610 77368-206 77389-530
1-Bromo-4,5-dichloro-2-isopropoxybenzene ≥97%
1-Bromo-4,5-dichloro-2-isopropoxybenzene

Precaución: for Research Use Only

Formula: C₉H₉BrCl₂O
MW: 283.9771 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761706
CAS Number: 2807448-47-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR