1,2-Dichloro-4-ethoxy-5-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

64293-500MG 64293-1G 64293-25G 64293-5G
77332-138EA 301.79 USD
77332-138 77356-518 77389-528 77380-462
1,2-Dichloro-4-ethoxy-5-iodobenzene ≥97%
1,2-Dichloro-4-ethoxy-5-iodobenzene

Precaución: for Research Use Only

Formula: C₈H₇Cl₂IO
MW: 316.9510 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761705
CAS Number: 2384268-69-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR