4,5-Dichloro-2-ethoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

64284-500MG 64284-1G 64284-5G
77337-028EA 405.82 USD
77337-028 77356-514 77380-458
4,5-Dichloro-2-ethoxybenzoic acid ≥97%
4,5-Dichloro-2-ethoxybenzoic acid

Precaución: for Research Use Only

Formula: C₉H₈Cl₂O₃
MW: 235.0640 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28580824
CAS Number: 1368036-11-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR