1-Bromo-4,5-dichloro-2-ethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

64281-500MG 64281-1G 64281-5G 64281-25G
77332-132EA 161.71 USD
77332-132 77345-924 77368-202 77389-522
1-Bromo-4,5-dichloro-2-ethoxybenzene ≥97%
1-Bromo-4,5-dichloro-2-ethoxybenzene

Precaución: for Research Use Only

Formula: C₈H₇BrCl₂O
MW: 269.9505 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761699
CAS Number: 2169321-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR