3-(Benzyloxy)-4,5-difluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

64256-500MG 64256-1G 64256-5G 64256-25G
77332-126EA 257.5 USD
77332-126 77356-506 77368-196 77389-516
3-(Benzyloxy)-4,5-difluorobenzaldehyde ≥97%
3-(Benzyloxy)-4,5-difluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₀F₂O₂
MW: 248.2248 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761683
CAS Number: 2764728-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR