3-(Benzyloxy)-4,5-difluorobenzoic acid ≥95%

Supplier: AOBChem USA

Warning

64253-500MG 64253-1G 64253-5G 64253-25G
77332-124EA 257.5 USD
77332-124 77356-504 77368-194 77389-514
3-(Benzyloxy)-4,5-difluorobenzoic acid ≥95%
3-(Benzyloxy)-4,5-difluorobenzoic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₀F₂O₃
MW: 264.2242 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761680
CAS Number: 2764728-25-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR