1-(2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)-2,2,2-trifluoroethanone ≥97%

Supplier: AOBChem USA

Warning

64245-500MG 64245-1G 64245-5G 64245-25G
77332-122EA 191.58 USD
77332-122 77356-502 77368-192 77389-512
1-(2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)-2,2,2-trifluoroethanone ≥97%
1-(2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)-2,2,2-trifluoroethanone

Precaución: for Research Use Only

Formula: C₁₀H₇F₃O₃
MW: 232.1559 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07380764
CAS Number: 122243-34-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR