2,2,2-Trifluoro-1-(4-isobutylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

64225-500MG 64225-1G 64225-5G 64225-25G
77332-120EA 150.38 USD
77332-120 77345-400 77368-190 77389-510
2,2,2-Trifluoro-1-(4-isobutylphenyl)ethanone ≥95%
2,2,2-Trifluoro-1-(4-isobutylphenyl)ethanone

Precaución: for Research Use Only

Formula: C₁₂H₁₃F₃O
MW: 230.2262 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07380763
CAS Number: 118569-68-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR