4'-Butyl-2,2,2-trifluoroacetophenone (carbonate ionophore IV) ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 4'-Butyl-2,2,2-trifluoroacetophenone, 1-(4-Butylphenyl)-2,2,2-trifluoroethanone

64223-500MG 64223-1G 64223-5G 64223-25G
77332-118EA 86.52 USD
77332-118 77342-270 77368-188 77380-456
4'-Butyl-2,2,2-trifluoroacetophenone (carbonate ionophore IV) ≥95%
4'-Butyl-2,2,2-trifluoroacetophenone (carbonate ionophore IV)

Precaución: for Research Use Only

Formula: C₁₂H₁₃F₃O
MW: 230.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00214157
CAS Number: 40739-44-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR