3,4-Difluoro-5-isopropoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

64204-500MG 64204-1G 64204-5G 64204-25G
77332-114EA 161.71 USD
77332-114 77345-922 77368-184 77389-506
3,4-Difluoro-5-isopropoxybenzaldehyde ≥97%
3,4-Difluoro-5-isopropoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₀F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD34761673
CAS Number: 1236364-47-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR