5-Bromo-1,2-difluoro-3-isopropoxybenzene ≥97%

Supplier: AOBChem USA

Warning

64199-1G 64199-5G 64199-25G
77341-350EA 82.4 USD
77341-350 77368-178 77380-454
5-Bromo-1,2-difluoro-3-isopropoxybenzene ≥97%
5-Bromo-1,2-difluoro-3-isopropoxybenzene

Precaución: for Research Use Only

Formula: C₉H₉BrF₂O
MW: 251.0679 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18917880
CAS Number: 1309932-71-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR