2,5-Difluoroisonicotinaldehyde ≥97%

Supplier: AOBChem USA

Warning

64169-1G 64169-5G 64169-25G
77341-884EA 106.09 USD
77341-884 77368-176 77380-444
2,5-Difluoroisonicotinaldehyde ≥97%
2,5-Difluoroisonicotinaldehyde

Precaución: for Research Use Only

Formula: C₆H₃F₂NO
MW: 143.0909 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12756646
CAS Number: 1227602-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR