4'-Chloro-2,2,2,3'-tetrafluoroacetophenone ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 1-(4-Chloro-3-fluorophenyl)-2,2,2-trifluoroethanone

64154-500MG 64154-1G 64154-5G 64154-25G
77332-102EA 86.52 USD
77332-102 77342-268 77368-174 77380-442
4'-Chloro-2,2,2,3'-tetrafluoroacetophenone ≥95%
4'-Chloro-2,2,2,3'-tetrafluoroacetophenone

Precaución: for Research Use Only

Formula: C₈H₃ClF₄O
MW: 226.55 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01319997
CAS Number: 845823-15-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR