1-(2,3-Difluoro-6-hydroxyphenyl)-2,2,2-trifluoroethanone ≥97%

Supplier: AOBChem USA

Warning

64146-500MG 64146-1G
77340-492EA 1267.93 USD
77340-492 77356-486
1-(2,3-Difluoro-6-hydroxyphenyl)-2,2,2-trifluoroethanone ≥97%
1-(2,3-Difluoro-6-hydroxyphenyl)-2,2,2-trifluoroethanone

Precaución: for Research Use Only

Formula: C₈H₃F₅O₂
Storage Temperature: Ambient
MDL Number: MFCD34761651
CAS Number: 1523219-35-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR