1-(2,4-Difluoro-3-methylphenyl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

64108-500MG 64108-1G 64108-5G
77332-096EA 342.99 USD
77332-096 77356-480 77380-434
1-(2,4-Difluoro-3-methylphenyl)-2,2,2-trifluoroethanone ≥95%
1-(2,4-Difluoro-3-methylphenyl)-2,2,2-trifluoroethanone

Precaución: for Research Use Only

Formula: C₉H₅F₅O
MW: 224.1274 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761648
CAS Number: 1509358-05-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR