1-(2,4-Difluoro-5-methylphenyl)-2,2,2-trifluoroethanone ≥97%

Supplier: AOBChem USA

Warning

64107-500MG 64107-1G 64107-5G 64107-25G
77332-094EA 202.91 USD
77332-094 77356-478 77368-170 77389-496
1-(2,4-Difluoro-5-methylphenyl)-2,2,2-trifluoroethanone ≥97%
1-(2,4-Difluoro-5-methylphenyl)-2,2,2-trifluoroethanone

Precaución: for Research Use Only

Formula: C₉H₅F₅O
MW: 224.1274 g/mol
Storage Temperature: Ambient
CAS Number: 1522817-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR