1-(3,4-Difluoro-2-methylphenyl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

64103-500MG 64103-1G 64103-5G
77332-088EA 338.87 USD
77332-088 77356-472 77380-428
1-(3,4-Difluoro-2-methylphenyl)-2,2,2-trifluoroethanone ≥95%
1-(3,4-Difluoro-2-methylphenyl)-2,2,2-trifluoroethanone

Precaución: for Research Use Only

Formula: C₉H₅F₅O
MW: 224.1274 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28131746
CAS Number: 1823320-28-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR