4'-Chloro-2'-methyl-2,2,2-trifluoroacetophenone ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 1-(4-Chloro-2-methylphenyl)-2,2,2-trifluoroethanone

64096-1G 64096-5G 64096-25G
77342-028EA 111.24 USD
77342-028 77368-168 77380-426
4'-Chloro-2'-methyl-2,2,2-trifluoroacetophenone ≥95%
4'-Chloro-2'-methyl-2,2,2-trifluoroacetophenone

Precaución: for Research Use Only

Formula: C₉H₆ClF₃O
MW: 222.60 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06201807
CAS Number: 845823-13-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR