1-(3-Chloro-2-methylphenyl)-2,2,2-trifluoroethanone ≥97%
Supplier: AOBChem USA
|
Warning
|
64094-500MG
64094-1G
64094-5G
64094-25G
77332-086EA
96.82
USD
77332-086
77342-646
77368-166
77380-424
1-(3-Chloro-2-methylphenyl)-2,2,2-trifluoroethanone ≥97%
1-(3-Chloro-2-methylphenyl)-2,2,2-trifluoroethanone
Precaución: for Research Use Only
Formula:
C₉H₆ClF₃O MW: 222.5915 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11113549 CAS Number: 1256467-21-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...