1-(3-Chloro-5-methylphenyl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

64092-500MG 64092-1G 64092-25G 64092-5G
77332-084EA 307.97 USD
77332-084 77356-470 77389-494 77380-422
1-(3-Chloro-5-methylphenyl)-2,2,2-trifluoroethanone ≥95%
1-(3-Chloro-5-methylphenyl)-2,2,2-trifluoroethanone

Precaución: for Research Use Only

Formula: C₉H₆ClF₃O
MW: 222.5915 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11556171
CAS Number: 1256479-46-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR