1-(2-Chloro-5-methylphenyl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

64091-500MG 64091-1G 64091-25G 64091-5G
77332-082EA 307.97 USD
77332-082 77356-468 77389-492 77380-420
1-(2-Chloro-5-methylphenyl)-2,2,2-trifluoroethanone ≥95%
1-(2-Chloro-5-methylphenyl)-2,2,2-trifluoroethanone

Precaución: for Research Use Only

Formula: C₉H₆ClF₃O
MW: 222.5915 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28105366
CAS Number: 1822673-79-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR