1-(2-Chloro-4-methylphenyl)-2,2,2-trifluoroethanone ≥97%

Supplier: AOBChem USA

Warning

64090-500MG 64090-1G 64090-25G 64090-5G
77332-080EA 307.97 USD
77332-080 77356-466 77389-490 77380-418
1-(2-Chloro-4-methylphenyl)-2,2,2-trifluoroethanone ≥97%
1-(2-Chloro-4-methylphenyl)-2,2,2-trifluoroethanone

Precaución: for Research Use Only

Formula: C₉H₆ClF₃O
MW: 222.5915 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28105250
CAS Number: 1824052-34-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR