2'-Methoxy-2,2,2,6'-tetrafluoroacetophenone ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-(2-Fluoro-6-methoxyphenyl)-2,2,2-trifluoroethan-1-one, 3-Fluoro-2-(trifluoroacetyl)anisole, 2,2,2-Trifluoro-1-(2-fluoro-6-methoxyphenyl)ethanone

64088-500MG 64088-1G 64088-5G 64088-25G
77332-078EA 117.42 USD
77332-078 77343-536 77368-164 77380-416
2'-Methoxy-2,2,2,6'-tetrafluoroacetophenone ≥97%
2'-Methoxy-2,2,2,6'-tetrafluoroacetophenone

Precaución: for Research Use Only

Formula: C₉H₆F₄O₂
MW: 222.136 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12922640
CAS Number: 1208078-28-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR