2,2,2-Trifluoro-1-(3-fluoro-5-methoxyphenyl)ethanone ≥97%
Supplier: AOBChem USA
|
Warning
|
64085-500MG
64085-1G
64085-5G
64085-25G
77332-076EA
237.93
USD
77332-076
77356-464
77368-162
77389-488
2,2,2-Trifluoro-1-(3-fluoro-5-methoxyphenyl)ethanone ≥97%
2,2,2-Trifluoro-1-(3-fluoro-5-methoxyphenyl)ethanone
Precaución: for Research Use Only
Formula:
C₉H₆F₄O₂ MW: 222.1363 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11556193 CAS Number: 1256482-19-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...